Assay Details
| Gene Symbol: LIN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005273102.3 | XP_005273159.1 | 1 | 42 | 81 | |
| XM_017001084.1 | XP_016856573.1 | 1 | 45 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005273102.3, XM_017001084.1 transcripts.
Set Membership: |
|

LIN9
Genomic Map