Assay Details
| Gene Symbol: ZNF496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011544315.2 | XP_011542617.1 | 12 | 3664 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF496| Gene Symbol: ZNF496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011544315.2 | XP_011542617.1 | 12 | 3664 | 64 | |
Set Membership: |
|