Assay Details
| Gene Symbol: NKIRAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001349.2 | NP_001001349.1 | 4 | 1089 | 104 | |
| NM_001144927.1 | NP_001138399.1 | 4 | 1193 | 104 | ||
| NM_001144928.1 | NP_001138400.1 | 4 | 1006 | 104 | ||
| NM_001144929.1 | NP_001138401.1 | 5 | 1077 | 104 | ||
| NM_017595.5 | NP_060065.2 | 4 | 1174 | 104 | ||
| XM_005257251.1 | XP_005257308.1 | 4 | 1201 | 104 | ||
| GenBank mRNA | AK027265.1 | - | 5 | 1046 | 104 | |
| AK054571.1 | - | 4 | 1201 | 104 | ||
| AK297975.1 | - | 4 | 974 | 104 | ||
| AL137682.1 | - | 2 | 2914 | 104 | ||
| BC007316.2 | - | 4 | 1126 | 104 | ||
| BC007450.2 | - | 4 | 1082 | 3028708 | 104 | |
| BC063498.1 | - | 4 | 1645 | 5248950 | 104 | |
More Information
Set Membership: |
|

NKIRAS2
Genomic Map