Assay Details
| Gene Symbol: GOSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321134.1 | NP_001308063.1 | 5 | 1205 | 95 | |
| NM_004287.4 | NP_004278.2 | 6 | 1298 | 95 | ||
| XM_005257844.3 | XP_005257901.1 | 5 | 1166 | 95 | ||
| XM_017025385.1 | XP_016880874.1 | 6 | 1304 | 95 | ||
| XM_017025389.1 | XP_016880878.1 | 6 | 1314 | 95 | ||
| XM_017025390.1 | XP_016880879.1 | 6 | 1659 | 95 | ||
| XM_017025393.1 | XP_016880882.1 | 5 | 1163 | 95 | ||
| XM_017025395.1 | XP_016880884.1 | 5 | 1514 | 95 | ||
More Information
Set Membership: |
|

GOSR2
Genomic Map