Assay Details
| Gene Symbol: LOC101927571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110782.1 | 2 | 84 | |||
| GenBank mRNA | AA928204.1 | - | 1 | 217 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927571| Gene Symbol: LOC101927571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110782.1 | 2 | 84 | |||
| GenBank mRNA | AA928204.1 | - | 1 | 217 | 84 | |
Set Membership: |
|