Assay Details
| Gene Symbol: PIAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324046.1 | NP_001310975.1 | 16 | 3881 | 92 | |
| NM_001324047.1 | NP_001310976.1 | 15 | 3821 | 92 | ||
| NM_001324048.1 | NP_001310977.1 | 16 | 3907 | 92 | ||
| NM_001324049.1 | NP_001310978.1 | 17 | 4030 | 92 | ||
| NM_004671.4 | NP_004662.2 | 14 | 3709 | 92 | ||
| NR_136684.1 | 13 | 3624 | 92 | |||
| XM_005258377.3 | XP_005258434.1 | 14 | 3610 | 92 | ||
| XM_005258379.3 | XP_005258436.1 | 14 | 3727 | 92 | ||
| XM_006722571.3 | XP_006722634.1 | 15 | 3695 | 92 | ||
| XM_006722572.3 | XP_006722635.1 | 15 | 3692 | 92 | ||
| XM_011526258.2 | XP_011524560.1 | 14 | 3775 | 92 | ||
| XM_011526259.2 | XP_011524561.1 | 15 | 3626 | 92 | ||
| XM_017026068.1 | XP_016881557.1 | 17 | 4011 | 92 | ||
| XM_017026069.1 | XP_016881558.1 | 15 | 3741 | 92 | ||
| XM_017026070.1 | XP_016881559.1 | 17 | 4008 | 92 | ||
More Information
Set Membership: |
|

PIAS2
Genomic Map