Assay Details
| Gene Symbol: ZBTB7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017025605.1 | XP_016881094.1 | 1 | 1726 | 89 | |
| XM_017025608.1 | XP_016881097.1 | 1 | 1726 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017025605.1, XM_017025608.1 transcripts.
Set Membership: |
|

ZBTB7C
Genomic Map