Assay Details
| Gene Symbol: ZBTB7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005259570.4 | XP_005259627.3 | 1 | 530 | 171 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005259570.4 transcripts.
Set Membership: |
|

ZBTB7A
Genomic Map