Assay Details
| Gene Symbol: NTF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011527009.2 | XP_011525311.1 | 1 | 180 | 105 | |
| XM_011527010.2 | XP_011525312.1 | 1 | 180 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011527009.2, XM_011527010.2 transcripts.
Set Membership: |
|

NTF4
Genomic Map