Assay Details
| Gene Symbol: ZNF480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011526465.2 | XP_011524767.1 | 1 | 135 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011526465.2 transcripts.
Set Membership: |
|

ZNF480
Genomic Map