Assay Details
| Gene Symbol: MAP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006718556.3 | XP_006718619.1 | 1 | 323 | 81 | |
| XM_011545039.2 | XP_011543341.1 | 1 | 324 | 81 | ||
| XM_017017755.1 | XP_016873244.1 | 1 | 223 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006718556.3, XM_011545039.2, XM_017017755.1 transcripts.
Set Membership: |
|

MAP6
Genomic Map