Assay Details
| Gene Symbol: EFNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001405.3 | NP_001396.2 | 4 | 1520 | 122 | |
| XM_017026449.1 | XP_016881938.1 | 5 | 3325 | 122 | ||
| XM_017026450.1 | XP_016881939.1 | 5 | 2209 | 122 | ||
More Information
Set Membership: |
|

EFNA2
Genomic Map