Assay Details
| Gene Symbol: ACER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011527673.2 | XP_011525975.1 | 1 - 2 | 194 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011527673.2 transcripts.
Set Membership: |
|

ACER1
Genomic Map