Assay Details
| Gene Symbol: SRP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003133.5 | NP_003124.1 | 2 - 3 | 266 | 111 | |
| GenBank mRNA | AF070649.1 | - | 2 - 3 | 245 | 111 | |
| AK289595.1 | - | 2 - 3 | 231 | 111 | ||
| BC008443.1 | - | 2 - 3 | 250 | 4102222 | 111 | |
| BC015094.1 | - | 1 | 237 | 3930047 | 111 | |
| BC022415.1 | - | 1 | 232 | 4243562 | 111 | |
| BC066957.1 | - | 2 - 3 | 253 | 5263635 | 111 | |
| DA983529.1 | - | 2 - 3 | 111 | |||
| U20998.1 | - | 2 - 3 | 244 | 111 | ||
More Information
Set Membership: |
|

SRP9
Genomic Map