Assay Details
| Gene Symbol: BIRC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011533005.2 | XP_011531307.1 | 58 | 13447 | 80 | |
| XM_017004560.1 | XP_016860049.1 | 58 | 13429 | 80 | ||
More Information
Set Membership: |
|

BIRC6
Genomic Map