Assay Details
| Gene Symbol: RGPD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017004843.1 | XP_016860332.1 | 21 | 5434 | 87 | |
| Gene Symbol: RGPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011532850.1 | XP_011531152.1 | 22 | 5486 | 87 | |
| Gene Symbol: RGPD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017004848.1 | XP_016860337.1 | 25 | 5784 | 87 | |
| Gene Symbol: RGPD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017005081.1 | XP_016860570.1 | 21 | 5425 | 87 | |
| Gene Symbol: RGPD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017004825.1 | XP_016860314.1 | 21 | 5507 | 87 | |
More Information
Set Membership: |
|

RGPD6
Genomic Map