Assay Details
| Gene Symbol: ARHGEF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005263681.1 | XP_005263738.1 | 1 - 2 | 79 | 84 | |
| XM_005263685.3 | XP_005263742.1 | 1 - 2 | 79 | 84 | ||
| XM_011511274.2 | XP_011509576.1 | 1 - 2 | 79 | 84 | ||
| XM_017004231.1 | XP_016859720.1 | 1 - 2 | 79 | 84 | ||
More Information
Set Membership: |
|

ARHGEF4
Genomic Map