Assay Details
| Gene Symbol: ACYP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320589.1 | NP_001307518.1 | 1 | 96 | 86 | |
| NM_001320590.1 | NP_001307519.1 | 1 | 96 | 86 | ||
| NM_138448.3 | NP_612457.1 | 1 | 96 | 86 | ||
| GenBank mRNA | BF739886.1 | - | 1 | 96 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001320589.1, NM_001320590.1, NM_138448.3 transcripts.
Set Membership: |
|

ACYP2
Genomic Map