Assay Details
| Gene Symbol: DNAJB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300914.1 | NP_001287843.1 | 2 | 581 | 78 | |
| NM_001313964.1 | NP_001300893.1 | 3 | 507 | 78 | ||
| NM_006145.2 | NP_006136.1 | 2 | 730 | 78 | ||
| XM_006722733.1 | XP_006722796.1 | 3 | 520 | 78 | ||
| XM_006722734.3 | XP_006722797.1 | 3 | 944 | 78 | ||
| XM_011527956.2 | XP_011526258.1 | 3 | 499 | 78 | ||
| GenBank mRNA | AK092989.1 | - | 1 | 133 | 78 | |
| AK301817.1 | - | 3 | 422 | 78 | ||
| AK312624.1 | - | 2 | 591 | 78 | ||
| BC002352.2 | - | 2 | 554 | 2821008 | 78 | |
| BC019827.1 | - | 2 | 544 | 4152608 | 78 | |
| CR536485.1 | - | 2 | 78 | |||
| CR541677.1 | - | 2 | 520 | 78 | ||
| D49547.1 | - | 2 | 560 | 78 | ||
More Information
Set Membership: |
|

DNAJB1
Genomic Map