Assay Details
| Gene Symbol: AHCY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011528658.2 | XP_011526960.2 | 11 | 3214 | 95 | |
| XM_017027709.1 | XP_016883198.1 | 11 | 3192 | 95 | ||
More Information
Set Membership: |
|

AHCY
Genomic Map