Assay Details
| Gene Symbol: NCR3LG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011520072.2 | XP_011518374.1 | 1 | 1179 | 110 | |
| XM_011520073.2 | XP_011518375.1 | 1 | 1179 | 110 | ||
| XM_011520075.2 | XP_011518377.1 | 1 | 1179 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011520072.2, XM_011520073.2, XM_011520075.2 transcripts.
Set Membership: |
|

NCR3LG1
Genomic Map