Assay Details
Gene Symbol: BRWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011529611.1 | XP_011527913.1 | 41 | 9579 | 117 | |
XM_011529612.2 | XP_011527914.1 | 35 | 8586 | 117 | ||
XM_011529613.1 | XP_011527915.1 | 20 | 6685 | 117 | ||
XM_017028373.1 | XP_016883862.1 | 43 | 9495 | 117 | ||
XM_017028374.1 | XP_016883863.1 | 18 | 8325 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
