Assay Details
| Gene Symbol: ETS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017028290.1 | XP_016883779.1 | 1 | 947 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017028290.1 transcripts.
Set Membership: |
|

ETS2
Genomic Map