Assay Details
| Gene Symbol: UFD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK315998.1 | - | 11 | 1279 | 76 | |
| BX648483.1 | - | 11 | 1260 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UFD1L| Gene Symbol: UFD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK315998.1 | - | 11 | 1279 | 76 | |
| BX648483.1 | - | 11 | 1260 | 76 | ||
Set Membership: |
|