Assay Details
| Gene Symbol: RFPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017028902.1 | XP_016884391.1 | 1 - 2 | 513 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017028902.1 transcripts.
Set Membership: |
|

RFPL1
Genomic Map