Assay Details
| Gene Symbol: TEF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB051442.1 | - | 1 | 5176 | 90 | |
| Gene Symbol: LOC100129648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK097637.1 | - | 1 | 90 | ||
More Information
Set Membership: |
|

TEF
Genomic Map