Assay Details
| Gene Symbol: PRPF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004698.2 | NP_004689.1 | 16 | 2213 | 107 | |
| XM_011510129.1 | XP_011508431.1 | 17 | 2255 | 107 | ||
| XM_011510130.2 | XP_011508432.1 | 13 | 2775 | 107 | ||
| XM_017002790.1 | XP_016858279.1 | 13 | 1649 | 107 | ||
| GenBank mRNA | AF001947.1 | - | 15 | 2106 | 107 | |
| AF016370.1 | - | 16 | 2123 | 107 | ||
| AK222825.1 | - | 16 | 2139 | 107 | ||
| AK297139.1 | - | 16 | 2097 | 107 | ||
| AK297265.1 | - | 15 | 1992 | 107 | ||
| BC000184.2 | - | 15 | 2100 | 3351519 | 107 | |
| BC001954.1 | - | 15 | 2108 | 2905308 | 107 | |
| BQ002848.1 | - | 1 | 107 | |||
More Information
Set Membership: |
|

PRPF3
Genomic Map