Assay Details
| Gene Symbol: MBNL1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027037.1 | 2 | 3965 | 140 | ||
| NR_027038.1 | 2 | 3976 | 140 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MBNL1-AS1| Gene Symbol: MBNL1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027037.1 | 2 | 3965 | 140 | ||
| NR_027038.1 | 2 | 3976 | 140 | |||
Set Membership: |
|