Assay Details
| Gene Symbol: UBXN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011512670.2 | XP_011510972.1 | 11 | 4899 | 105 | |
| XM_011512671.2 | XP_011510973.1 | 10 | 4756 | 105 | ||
More Information
Set Membership: |
|

UBXN7
Genomic Map