Assay Details
| Gene Symbol: NKIRAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005265076.3 | XP_005265133.1 | 5 | 3783 | 119 | |
| XM_005265077.4 | XP_005265134.1 | 4 | 3751 | 119 | ||
| XM_005265078.3 | XP_005265135.1 | 5 | 3735 | 119 | ||
More Information
Set Membership: |
|

NKIRAS1
Genomic Map