Assay Details
Gene Symbol: FOXL2NB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040061.2 | NP_001035150.1 | 3 | 3851 | 104 | |
XM_005247443.3 | XP_005247500.1 | 4 | 3937 | 104 | ||
GenBank mRNA | AK125319.1 | - | 3 | 3851 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
