Assay Details
| Gene Symbol: TP63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114978.1 | NP_001108450.1 | 13 | 2995 | 116 | |
| NM_001114980.1 | NP_001108452.1 | 12 | 2859 | 116 | ||
| NM_001114981.1 | NP_001108453.1 | 11 | 2765 | 116 | ||
| NM_003722.4 | NP_003713.3 | 14 | 3089 | 116 | ||
| XM_005247844.4 | XP_005247901.1 | 14 | 3252 | 116 | ||
| XM_011513251.2 | XP_011511553.1 | 15 | 3130 | 116 | ||
| XM_011513252.2 | XP_011511554.1 | 14 | 3062 | 116 | ||
| GenBank mRNA | AF091627.1 | - | 12 | 2941 | 116 | |
| BC039815.1 | - | 14 | 3089 | 5552611 | 116 | |
| GQ202690.1 | - | 10 | 2618 | 116 | ||
| Y16961.1 | - | 14 | 3027 | 116 | ||
More Information
Set Membership: |
|

TP63
Genomic Map