Assay Details
| Gene Symbol: MOGAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025098.3 | NP_079374.2 | 3 | 390 | 99 | |
| XM_011545267.1 | XP_011543569.1 | 3 | 390 | 99 | ||
| GenBank mRNA | AK026297.1 | - | 3 | 389 | 99 | |
| AK128620.1 | - | 2 | 847 | 99 | ||
| AK291998.1 | - | 3 | 366 | 99 | ||
| AY157608.1 | - | 3 | 320 | 99 | ||
| BC103876.1 | - | 3 | 320 | 40004273 | 99 | |
| BC103877.1 | - | 3 | 320 | 40004282 | 99 | |
| BC103878.1 | - | 3 | 320 | 40004283 | 99 | |
| DA922421.1 | - | 3 | 391 | 99 | ||
More Information
Set Membership: |
|

MOGAT2
Genomic Map