Assay Details
| Gene Symbol: FAM160A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109977.1 | NP_001103447.1 | 1 | 141 | 107 | |
| XM_005263199.1 | XP_005263256.1 | 1 | 141 | 107 | ||
| XM_011532221.2 | XP_011530523.1 | 1 | 244 | 107 | ||
| XM_011532222.2 | XP_011530524.1 | 1 | 244 | 107 | ||
| XM_011532223.2 | XP_011530525.1 | 1 | 244 | 107 | ||
| GenBank mRNA | DA722299.1 | - | 1 | 141 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109977.1, XM_005263199.1, XM_011532221.2, XM_011532222.2, XM_011532223.2 transcripts.
Set Membership: |
|

FAM160A1
Genomic Map