Assay Details
| Gene Symbol: DEPDC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011543510.2 | XP_011541812.1 | 8 | 1725 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DEPDC1B| Gene Symbol: DEPDC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011543510.2 | XP_011541812.1 | 8 | 1725 | 108 | |
Set Membership: |
|