Assay Details
| Gene Symbol: LOC107986353 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017010107.1 | XP_016865596.1 | 1 | 1625 | 93 | |
| Gene Symbol: LOC107986351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017010104.1 | XP_016865593.1 | 1 | 93 | ||
| Gene Symbol: LOC107986352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017010106.1 | XP_016865595.1 | 1 | 1625 | 93 | |
| Gene Symbol: LOC107986354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017010108.1 | XP_016865597.1 | 1 | 1626 | 93 | |
| Gene Symbol: LOC441081 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073404.1 | 1 | 93 | |||
| Gene Symbol: LOC728488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK296222.1 | - | 2 | 93 | ||
| AK311627.1 | - | 2 | 93 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017010106.1, XM_017010107.1, XM_017010108.1 transcripts.
Set Membership: |
|

LOC107986353
Genomic Map