Assay Details
| Gene Symbol: COL4A3BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130105.1 | NP_001123577.1 | 19 | 5278 | 122 | |
| NM_005713.2 | NP_005704.1 | 18 | 5179 | 122 | ||
| NM_031361.2 | NP_112729.1 | 17 | 5101 | 122 | ||
| XM_011543090.2 | XP_011541392.1 | 17 | 5042 | 122 | ||
| XM_011543091.2 | XP_011541393.1 | 16 | 4964 | 122 | ||
| GenBank mRNA | AK125387.1 | - | 1 | 523 | 122 | |
More Information
Set Membership: |
|

COL4A3BP
Genomic Map