Assay Details
| Gene Symbol: CCNH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199189.1 | NP_001186118.1 | 1 | 408 | 66 | |
| XM_005248629.4 | XP_005248686.1 | 1 | 470 | 66 | ||
| XM_017010014.1 | XP_016865503.1 | 1 | 434 | 66 | ||
| GenBank mRNA | BX537673.1 | - | 1 | 409 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001199189.1, XM_005248629.4, XM_017010014.1 transcripts.
Set Membership: |
|

CCNH
Genomic Map