Assay Details
| Gene Symbol: LOC105379143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134250.1 | 3 | 332 | 114 | ||
| GenBank mRNA | DB288884.1 | - | 3 | 332 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105379143| Gene Symbol: LOC105379143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134250.1 | 3 | 332 | 114 | ||
| GenBank mRNA | DB288884.1 | - | 3 | 332 | 114 | |
Set Membership: |
|