Assay Details
| Gene Symbol: P4HA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005272116.4 | XP_005272173.1 | 15 | 3030 | 152 | |
| XM_005272117.4 | XP_005272174.1 | 16 | 3188 | 152 | ||
| XM_005272118.4 | XP_005272175.1 | 16 | 3317 | 152 | ||
| XM_005272119.4 | XP_005272176.1 | 16 | 3507 | 152 | ||
| XM_005272120.4 | XP_005272177.1 | 16 | 3287 | 152 | ||
| XM_006714728.3 | XP_006714791.1 | 16 | 3564 | 152 | ||
| XM_006714729.3 | XP_006714792.1 | 16 | 3182 | 152 | ||
| XM_006714730.3 | XP_006714793.1 | 16 | 3558 | 152 | ||
| XM_011543705.2 | XP_011542007.1 | 13 | 2779 | 152 | ||
| XM_017010011.1 | XP_016865500.1 | 16 | 3281 | 152 | ||
| XM_017010012.1 | XP_016865501.1 | 14 | 2847 | 152 | ||
More Information
Set Membership: |
|

P4HA2
Genomic Map