Assay Details
| Gene Symbol: TROVE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006711495.3 | XP_006711558.1 | 10 | 8664 | 112 | |
| XM_006711496.3 | XP_006711559.1 | 11 | 8840 | 112 | ||
| XM_006711497.3 | XP_006711560.1 | 10 | 8774 | 112 | ||
| XM_011509922.2 | XP_011508224.1 | 9 | 8173 | 112 | ||
| XM_017002180.1 | XP_016857669.1 | 10 | 8473 | 112 | ||
| XM_017002181.1 | XP_016857670.1 | 10 | 8702 | 112 | ||
| XM_017002182.1 | XP_016857671.1 | 9 | 8407 | 112 | ||
| XM_017002183.1 | XP_016857672.1 | 7 | 7744 | 112 | ||
More Information
Set Membership: |
|

TROVE2
Genomic Map