Assay Details
| Gene Symbol: KCTD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005273158.1 | XP_005273215.1 | 1 | 88 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005273158.1 transcripts.
Set Membership: |
|

KCTD3
Genomic Map