Assay Details
| Gene Symbol: LOC101927401 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011544350.2 | XP_011542652.1 | 6 - 7 | 1623 | 120 | |
| XM_011544351.2 | XP_011542653.1 | 2 - 3 | 776 | 120 | ||
| XM_011544352.2 | XP_011542654.1 | 2 - 3 | 692 | 120 | ||
| XM_017003091.1 | XP_016858580.1 | 5 - 6 | 1406 | 120 | ||
| XM_017003092.1 | XP_016858581.1 | 6 - 7 | 1683 | 120 | ||
More Information
Set Membership: |
|

LOC101927401
Genomic Map