Assay Details
| Gene Symbol: MTR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011544194.2 | XP_011542496.1 | 1 | 293 | 80 | |
| XM_017001329.1 | XP_016856818.1 | 1 | 293 | 80 | ||
| XM_017001330.1 | XP_016856819.1 | 1 | 293 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011544194.2, XM_017001329.1, XM_017001330.1 transcripts.
Set Membership: |
|

MTR
Genomic Map