Assay Details
| Gene Symbol: PCDHA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018902.4 | NP_061725.1 | 1 | 224 | 91 | |
| NM_031861.2 | NP_114067.1 | 1 | 224 | 91 | ||
| GenBank mRNA | BX111117.1 | - | 1 | 236 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_018902.4, NM_031861.2 transcripts.
Set Membership: |
|

PCDHA11
Genomic Map