Assay Details
| Gene Symbol: EBF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017009199.1 | XP_016864688.1 | 1 | 1124 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017009199.1 transcripts.
Set Membership: |
|

EBF1
Genomic Map