Assay Details
| Gene Symbol: LOC105377879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BI460215.1 | - | 1 - 2 | 290 | 86 | |
| CR745838.1 | - | 1 - 2 | 173 | 86 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105377879| Gene Symbol: LOC105377879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BI460215.1 | - | 1 - 2 | 290 | 86 | |
| CR745838.1 | - | 1 - 2 | 173 | 86 | ||
Set Membership: |
|