Assay Details
| Gene Symbol: MEMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301833.1 | NP_001288762.1 | 3 | 249 | 89 | |
| NM_001301852.1 | NP_001288781.1 | 2 | 171 | 89 | ||
| NM_015955.3 | NP_057039.1 | 2 | 538 | 89 | ||
| NR_126032.1 | 2 | 538 | 89 | |||
| NR_126034.1 | 2 | 171 | 89 | |||
| XM_011532892.2 | XP_011531194.1 | 3 | 186 | 89 | ||
| XM_017004255.1 | XP_016859744.1 | 3 | 159 | 89 | ||
| GenBank mRNA | AB041018.1 | - | 2 | 538 | 89 | |
| AB041019.1 | - | 2 | 187 | 89 | ||
| AF132961.1 | - | 2 | 236 | 89 | ||
| AF363446.1 | - | 2 | 455 | 89 | ||
| AF529368.1 | - | 2 | 121 | 89 | ||
| AK057760.1 | - | 2 | 160 | 89 | ||
| AK290753.1 | - | 3 | 205 | 89 | ||
| BC018733.1 | - | 2 | 503 | 4851517 | 89 | |
| BC070046.1 | - | 3 | 278 | 5273070 | 89 | |
| BC094681.1 | - | 2 | 142 | 6205399 | 89 | |
| BG033287.1 | - | 2 | 120 | 89 | ||
| BP248979.1 | - | 3 | 249 | 89 | ||
| CD250945.1 | - | 2 | 235 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001301852.1 transcripts.
Set Membership: |
|

MEMO1
Genomic Map