Assay Details
| Gene Symbol: RAET1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017010285.1 | XP_016865774.1 | 8 | 2677 | 94 | |
| XM_017010286.1 | XP_016865775.1 | 9 | 2596 | 94 | ||
| XM_017010287.1 | XP_016865776.1 | 8 | 2677 | 94 | ||
More Information
Set Membership: |
|

RAET1E
Genomic Map