Assay Details
| Gene Symbol: MTRF1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017010959.1 | XP_016866448.1 | 1 | 3389 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017010959.1 transcripts.
Set Membership: |
|

MTRF1L
Genomic Map